missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GDF-8 / Myostatin / MSTN Protein
Recombinant Protein
107.00 € - 7105.00 €
Specifications
Accession Number | NP_005250.1 |
---|---|
Gene ID (Entrez) | 2660 |
Name | GDF-8 / Myostatin / MSTN Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Less than or equal to 1.00 EUs/ug as determined by the LAL method. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15995799
|
enQuireBio™
QP5265-5UG |
5 μg |
107.00 €
5µg |
Estimated Shipment: 28-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15975799
|
enQuireBio™
QP5265-25UG |
25 μg |
252.00 €
25µg |
Estimated Shipment: 28-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15955799
|
enQuireBio™
QP5265-100UG |
100 μg |
940.00 €
100µg |
Estimated Shipment: 28-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15985799
|
enQuireBio™
QP5265-500UG |
500 μg |
4325.00 €
500µg |
Estimated Shipment: 28-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15965799
|
enQuireBio™
QP5265-1MG |
1 mg |
7105.00 €
1mg |
Estimated Shipment: 28-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
NP_005250.1 | |
GDF-8 / Myostatin / MSTN Protein | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Bioactivity of Myostatin Propeptide was confirmed by measuring inhibition of myostatin induced cytotoxicity in MCP-11 cells. Typical ED50 is around 1 ug/ml myostatin propeptide. | |
Human | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |
2660 | |
Research Use Only | |
MSTN | |
Recombinant Protein | |
E. coli | |
MNENSEQKEN VEKEGLCNAC TWRQNTKSSR IEAIKIQILS KLRLETAPNI SKDVIRQLLP KAPPLRELID QYDVQRDDSS DGSLEDDDYH ATTETIITMP TESDFLMQVD GKPKCCFFKF SSKIQYNKVV KAQLWIYLRP VETPTTVFVQ ILRLIKPMKD GTRYTGIRSL KLDMNPGTGI WQSIDVKTVL QNWLKQPESN LGIEIKALDE NGHDLAVTFP GPGEDGLNPF LEVKVTDTPK RSRR | |
Greater than 95% as determined by SDS-PAGE. |