missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GDF-8 / Myostatin / MSTN Protein
Recombinant Protein
Brand: enQuireBio™ QP5265-5ug
Additional Details : Weight : 0.01000kg
Specifications
NP_005250.1 | |
GDF-8 / Myostatin / MSTN Protein | |
Research Use Only | |
MSTN | |
Recombinant Protein | |
E. coli | |
MNENSEQKEN VEKEGLCNAC TWRQNTKSSR IEAIKIQILS KLRLETAPNI SKDVIRQLLP KAPPLRELID QYDVQRDDSS DGSLEDDDYH ATTETIITMP TESDFLMQVD GKPKCCFFKF SSKIQYNKVV KAQLWIYLRP VETPTTVFVQ ILRLIKPMKD GTRYTGIRSL KLDMNPGTGI WQSIDVKTVL QNWLKQPESN LGIEIKALDE NGHDLAVTFP GPGEDGLNPF LEVKVTDTPK RSRR | |
Greater than 95% as determined by SDS-PAGE. |
2660 | |
5 μg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Bioactivity of Myostatin Propeptide was confirmed by measuring inhibition of myostatin induced cytotoxicity in MCP-11 cells. Typical ED50 is around 1 ug/ml myostatin propeptide. | |
Human | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |