missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human GDNF Protein

Product Code. 18473352 Shop All Bio Techne Products
Click to view available options
Quantity:
1 mg
10 μg
100 μg
500 μg
Unit Size:
100µg
10µg
1mg
500µg
This item is not returnable. View return policy

Product Code. 18473352

Brand: Novus Biologicals™ NBP234888500ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Fully biologically active when compared to standard. The ED50 as determined by a cell proliferation assay using rat C6 cells is less than 0.1 ng/mL, corresponding to a specific activity of >5.0 x 107 IU/mg.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P39905, P39905, P39905
Concentration LYOPH
For Use With (Application) PAGE, Bioactivity
Formulation Lyophilized from a 0.2 um filtered concentrated solution in 1 x PBS, pH 7.4, with 0.05 % Tween-20.
Gene ID (Entrez) 2668
Molecular Weight (g/mol) 30.1 kDa
Quantity 500 μg
Storage Requirements Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles.
Endotoxin Concentration Less than 0.1 EU/ug of GDNF as determined by LAL method.
Gene Alias Astrocyte-derived trophic factor, ATF, ATF1, ATF2, glial cell derived neurotrophic factor, glial cell line derived neurotrophic factor, glial cell line-derived neurotrophic factor, HFB1-GDNF, hGDNF, HSCR3
Gene Symbol GDNF
Reconstitution Recommended to centrifuge prior to opening. Reconstitute in sterile distilled water or aqueous buffer containing 0.1% BSA to a concentration of 0.1-1.0mg/mL. Apportion stock solutions into working aliquots and store at <-20°C.
Research Category Apoptosis, Biologically Active Proteins, Cancer
Purity or Quality Grade >97% pure by SDS-PAGE and HPLC
Protein GDNF
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.