missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human Glutamate Receptor 3 GST (N-Term) Protein

Product Code. p-200051279 Shop All Bio Techne Products
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
This item is not returnable. View return policy

Product Code. 18326508

Brand: Novus Biologicals™ H00002892Q0125ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.
TRUSTED_SUSTAINABILITY

Especificaciones

For Use With (Application) Western Blot, ELISA, PAGE, Protein Array, Immunoaffinity Purification
Formulation 50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer.
Gene ID (Entrez) 2892
Molecular Weight (g/mol) 36.74 kDa
Quantity 25 μg
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Gene Alias AMPA 3, AMPA-selective glutamate receptor 3, GluA3, GluR-3, GLUR-C, GLUR-K3, glutamate receptor C, glutamate receptor subunit 3, glutamate receptor, ionotrophic, AMPA 3
Gene Symbol GRIA3
Research Category Neuronal Cell Markers, Neuroscience, Neurotransmission, Vision
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Protein Glutamate Receptor 3
Mostrar más Mostrar menos
Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado