missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Glycine N-acyltransferase-like protein 1
A DNA sequence encoding the Homo sapiens (Human) Glycine N-acyltransferase-like protein 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7893-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001207423.1 | |
Glycine N-acyltransferase-like protein 1 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
92292 | |
Greater than 80% as determined by SDS-PAGE. | |
Research Use Only | |
GLYATL1 | |
Recombinant Protein | |
E. coli | |
MILLNNSHKL LALYKSLARS IPESLKVYGS VYHINHGNPF NMEVLVDSWP EYQMVIIRPQ KQEMTDDMDS YTNVYRMFSK EPQKSEEVLK NCEIVNWKQR LQIQGLQESL GEGIRVATFS KSVKVEHSRA LLLVTEDILK LNASSKSKLG SWAETGHPDD EFESETPNFK YAQLDVSYSG LVNDNWKRGK NERSLHYIKR CIEDLPAACM LGPEGVPVSW VTMDPSCEVG MAYSMEKYRR TGNMARVMVR YMKYLRQKNI PFYISVLEEN EDSRRFVGQF GFFEASCEWH QWTCYPQNLV PF |