missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Glycodelin Protein

A DNA sequence encoding the Homo sapiens (Human) Glycodelin, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6462-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16033470

  • 505.00 € / 100µg
Estimated Shipment: 16-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001018058.1
Glycodelin Protein
Research Use Only
PAEP
Recombinant Protein
E. coli
MDIPQTKQDL ELPKLAGTWH SMAMATNNIS LMATLKAPLR VHITSLLPTP EDNLEIVLHR WENNSCVEKK VLGEKTENPK KFKINYTVAN EATLLDTDYD NFLFLCLQDT TTPIQSMMCQ YLARVLVEDD EIMQGFIRAF RPLPRHLWYL LDLKQMEEPC RF
Greater than 90% as determined by SDS-PAGE.
5047
100 μg
Not determined.
Full Length of Mature Protein
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers