missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Glycodelin Protein
A DNA sequence encoding the Homo sapiens (Human) Glycodelin, was expressed in the hosts and tags indicated.
203.00 € - 2285.00 €
Specifications
Accession Number | NP_001018058.1 |
---|---|
Gene ID (Entrez) | 5047 |
Name | Glycodelin Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16043470
|
enQuireBio™
QP6462-EC-10UG |
10 μg |
203.00 €
10µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16033470
|
enQuireBio™
QP6462-EC-100UG |
100 μg |
505.00 €
100µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16053470
|
enQuireBio™
QP6462-EC-1MG |
1 mg |
2285.00 €
1mg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16063470
|
enQuireBio™
QP6462-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16073470
|
enQuireBio™
QP6462-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16083470
|
enQuireBio™
QP6462-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001018058.1 | |
Glycodelin Protein | |
Not determined. | |
Full Length of Mature Protein | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
5047 | |
Research Use Only | |
PAEP | |
Recombinant Protein | |
E. coli | |
MDIPQTKQDL ELPKLAGTWH SMAMATNNIS LMATLKAPLR VHITSLLPTP EDNLEIVLHR WENNSCVEKK VLGEKTENPK KFKINYTVAN EATLLDTDYD NFLFLCLQDT TTPIQSMMCQ YLARVLVEDD EIMQGFIRAF RPLPRHLWYL LDLKQMEEPC RF | |
Greater than 90% as determined by SDS-PAGE. |