missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human GMPS His Protein

Product Code. 18047416 Shop All Bio Techne Products
Click to view available options
:
50μg; Unlabeled
This item is not returnable. View return policy

Product Code. 18047416

Brand: Novus Biologicals™ NBP19911450UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-693 of Human GMPS The Recombinant Human GMPS Protein is derived from E. coli. The Recombinant Human GMPS Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P49915
For Use With (Application) SDS-PAGE
Formulation Liquid. In 20mM Tris-HCl buffer (pH 8.0) containing 1mM DTT, 30% glycerol, 0.1M NaCl
Gene ID (Entrez) 8833
Molecular Weight (g/mol) 79.2kDa
Name GMPS Protein
Purification Method SDS-PAGE
Storage Requirements 4°C short term, −20°C long term. Avoid freeze-thaw cycles.
Cross Reactivity Human
Purity or Quality Grade >90%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.