missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HDHD1 Protein
A cDNA sequence encoding the HDHD1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12208-1mg
Additional Details : Weight : 0.01000kg
Specifications
8226 | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MAAPPQPVTH LIFDMDGLLL DTERLYSVVF QEICNRYDKK YSWDVKSLVM GKKALEAAQI IIDVLQLPMS KEELVEESQT KLKEVFPTAA LMPGAEKLII HLRKHGIPFA LATSSGSASF DMKTSRHKEF FSLFSHIVLG DDPEVQHGKP DPDIFLACAK RFSPPPAMEK CLVFEDAPNG VEAALAAGMQ VVMVPDGNLS RDLTTKATLV LNSLQDFQPE LFGLPSYE | |
Greater than 95.0% as determined by SDS-PAGE. |
HDHD1 Protein | |
Research Use Only | |
PUDP | |
Human | |
His | |
The HDHD1 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 20% glycerol,0.1M NaCl and 1mM DTT. |