missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HDHD1 Protein
A cDNA sequence encoding the HDHD1 was constructed and used to recombinantly synthesize the protein.
131.00 € - 3915.00 €
Specifications
Gene ID (Entrez) | 8226 |
---|---|
Name | HDHD1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | PUDP |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15959698
|
enQuireBio™
QP12208-5UG |
5 μg |
131.00 €
5µg |
Estimated Shipment: 28-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15949698
|
enQuireBio™
QP12208-20UG |
20 μg |
203.00 €
20µg |
Estimated Shipment: 28-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15939698
|
enQuireBio™
QP12208-1MG |
1 mg |
3915.00 €
1mg |
Estimated Shipment: 28-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
8226 | |
Research Use Only | |
PUDP | |
Human | |
His | |
The HDHD1 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 20% glycerol,0.1M NaCl and 1mM DTT. |
HDHD1 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MAAPPQPVTH LIFDMDGLLL DTERLYSVVF QEICNRYDKK YSWDVKSLVM GKKALEAAQI IIDVLQLPMS KEELVEESQT KLKEVFPTAA LMPGAEKLII HLRKHGIPFA LATSSGSASF DMKTSRHKEF FSLFSHIVLG DDPEVQHGKP DPDIFLACAK RFSPPPAMEK CLVFEDAPNG VEAALAAGMQ VVMVPDGNLS RDLTTKATLV LNSLQDFQPE LFGLPSYE | |
Greater than 95.0% as determined by SDS-PAGE. |