missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human HDHD1 Protein

A cDNA sequence encoding the HDHD1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12208-5ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15959698

  • 131.00 € / 5µg
Estimated Shipment: 28-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

8226
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MAAPPQPVTH LIFDMDGLLL DTERLYSVVF QEICNRYDKK YSWDVKSLVM GKKALEAAQI IIDVLQLPMS KEELVEESQT KLKEVFPTAA LMPGAEKLII HLRKHGIPFA LATSSGSASF DMKTSRHKEF FSLFSHIVLG DDPEVQHGKP DPDIFLACAK RFSPPPAMEK CLVFEDAPNG VEAALAAGMQ VVMVPDGNLS RDLTTKATLV LNSLQDFQPE LFGLPSYE
Greater than 95.0% as determined by SDS-PAGE.
HDHD1 Protein
Research Use Only
PUDP
Human
His
The HDHD1 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 20% glycerol,0.1M NaCl and 1mM DTT.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers