missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HNRNPL Protein
A DNA sequence encoding the Homo sapiens (Human) HNRNPL, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9182-ye-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001005335.1 | |
HNRNPL Protein | |
Research Use Only | |
HNRNPL | |
Recombinant Protein | |
Yeast | |
IRGLIDGVVE ADLVEALQEF GPISYVVVMP KKRQALVEFE DVLGACNAVN YAADNQIYIA GHPAFVNYST SQKISRPGDS DDSRSVNSVL LFTILNPIYS ITTDVLYTIC NPCGPVQRIV IFRKNGVQAM VEFDSVQSAQ RAKASLNGAD IYSGCCTLKI EYAKPTRLNV FKNDQDTWDY TNPNLSGQGD PGSNPNKRQR QPPLLGDHPA EYGGPHGGYH SHYHDEGYGP PPPHYEGRRM GPPVGGH | |
Greater than 90% as determined by SDS-PAGE. |
3191 | |
100 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |