missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human HSPB1-associated protein 1

A DNA sequence encoding the Homo sapiens (Human) HSPB1-associated protein 1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7985-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16061991

  • 2285.00 € / 1mg
Estimated Shipment: 05-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001307657.1
HSPB1-associated protein 1
Research Use Only
HSPBAP1
Recombinant Protein
E. coli
MAAGSEATTP VIVAAGAGGE EGEHVKPFKP EKAKEIIMSL QQPAIFCNMV FDWPARHWNA KYLSQVLHGK QIRFRMGMKS MSTVPQFETT CNYVEATLEE FLTWNCDQSS ISGPFRDYDH SKFWAYADYK YFVSLFEDKT DLFQDVKWSD FGFPGRNGQE STLWIGSLGA HTPCHLDSYG CNLVFQVQGR KRWHLFPPED TPFLYPTRIP YEESSVFSKI NVVNPDLKRF PQFRKAQRHA VTLSPGQVLF VPRHWWHYVE SIDPVTVSIN SWIELEEDHL ARVEEAITRM LVCALKTAEN PQNTRAWLNP TEVEETSHAV NCCYLNAAVS AFFDRCRTSE VVEIQALRTD GEHMKKEELN VCNHMEVGQT GSQNLTTGTD KPEAASPFGP DLVPVAQRSE EPPSERGGIF GSDGKDFVDK DGEHFGKLHC AKRQQIMSNS ENAIEEQIAS NTTTTPQTFI STDDLLDCLV NPQVTRIVAQ LLIQGRSL
Greater than 90% as determined by SDS-PAGE.
79663
1 mg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers