missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HSPB1-associated protein 1
A DNA sequence encoding the Homo sapiens (Human) HSPB1-associated protein 1, was expressed in the hosts and tags indicated.
160.00 € - 2285.00 €
Specifications
Accession Number | NP_001307657.1 |
---|---|
Gene ID (Entrez) | 79663 |
Name | HSPB1-associated protein 1 |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16041991
|
enQuireBio™
QP7985-EC-100UG |
100 μg |
505.00 €
100µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16061991
|
enQuireBio™
QP7985-EC-1MG |
1 mg |
2285.00 €
1mg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16051991
|
enQuireBio™
QP7985-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16071991
|
enQuireBio™
QP7985-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16081991
|
enQuireBio™
QP7985-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16091991
|
enQuireBio™
QP7985-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001307657.1 | |
HSPB1-associated protein 1 | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
79663 | |
Research Use Only | |
HSPBAP1 | |
Recombinant Protein | |
E. coli | |
MAAGSEATTP VIVAAGAGGE EGEHVKPFKP EKAKEIIMSL QQPAIFCNMV FDWPARHWNA KYLSQVLHGK QIRFRMGMKS MSTVPQFETT CNYVEATLEE FLTWNCDQSS ISGPFRDYDH SKFWAYADYK YFVSLFEDKT DLFQDVKWSD FGFPGRNGQE STLWIGSLGA HTPCHLDSYG CNLVFQVQGR KRWHLFPPED TPFLYPTRIP YEESSVFSKI NVVNPDLKRF PQFRKAQRHA VTLSPGQVLF VPRHWWHYVE SIDPVTVSIN SWIELEEDHL ARVEEAITRM LVCALKTAEN PQNTRAWLNP TEVEETSHAV NCCYLNAAVS AFFDRCRTSE VVEIQALRTD GEHMKKEELN VCNHMEVGQT GSQNLTTGTD KPEAASPFGP DLVPVAQRSE EPPSERGGIF GSDGKDFVDK DGEHFGKLHC AKRQQIMSNS ENAIEEQIAS NTTTTPQTFI STDDLLDCLV NPQVTRIVAQ LLIQGRSL | |
Greater than 90% as determined by SDS-PAGE. |