missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human IGF-I/IGF-1 Protein
Shop All Bio Techne Products
Click to view available options
Quantity:
100 μg
500 μg
Unit Size:
100µg
500µg
Description
IGF1 Protein is fully biologically active when compared to standard. The ED50 as determined by a cell proliferation assay using serum free human MCF-7 cells is less than 2 ng/mL, corresponding to a specific activity of > 5.0 x 10^5 IU/mg.
Specifications
Specifications
| Accession Number | P05019, P05019, P05019 |
| Concentration | LYOPH |
| For Use With (Application) | SDS-PAGE |
| Formulation | Lyophilized from a 0.2 um filtered concentrated solution in PBS, pH 7.0. |
| Gene ID (Entrez) | 3479 |
| Molecular Weight (g/mol) | M.W. Theoretical: 7.6 kDa |
| Quantity | 100 μg |
| Storage Requirements | Store at -20 to -70 C as supplied. After reconstitution, store at 2 to 8 C for 1 month and (at -20 to -70 C for long term storage. Avoid repeated freeze-thaw cycles.) |
| Endotoxin Concentration | Less than 0.01 EU/ug of IGF-I/IGF-1 AS determined by LAL method. |
| Gene Alias | IBP1, IGF1A, IGFI, IGF-I, IGF-IA, IGF-IB, insulin-like growth factor 1, insulin-like growth factor 1 (somatomedin C), insulin-like growth factor I, insulin-like growth factor IA, insulin-like growth factor IB, Mechano growth factor, MGF, somatomedin-C |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction