missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human IGSF8 Protein

A cDNA sequence encoding the IGSF8 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12389-1ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15960259

  • 131.00 € / 1µg
Estimated Shipment: 26-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

IGSF8 Protein
Research Use Only
Recombinant Protein
Baculovirus
ADLREVLVPE GPLYRVAGTA VSISCNVTGY EGPAQQNFEW FLYRPEAPDT ALGIVSTKDT QFSYAVFKSR VVAGEVQVQR LQGDAVVLKI ARLQAQDAGI YECHTPSTDT RYLGSYSGKV ELRVLPDVLQ VSAAPPGPRG RQAPTSPPRM TVHEGQELAL GCLARTSTQK HTHLAVSFGR SVPEAPVGRS TLQEVVGIRS DLAVEAGAPY AERLAAGELR LGKEGTDRYR MVVGGAQAGD AGTYHCTAAE WIQDPDGSWA QIAEKRAVLA HVDVQTLSSQ LAVTVGPGER RIGPGEPLEL LCNVSGALPP AGRHAAYSVG WEMAPAGAPG PGRLVAQLDT EGVGSLGPGY EGRHIAMEKV ASRTYRLRLE AARPGDAGTY RCLAKAYVRG SGTRLREAAS ARSRPLPVHV REEGVVLEAV AWLAGGTVYR GETASLLCNI SVRGGPPGLR LAASWWVERP EDGELSSVPA QLVGGVGQDG VAELGVRPGG GPVSVELVGP RSHRLRLHSL GPEDEGVYHC APSAWVQHAD YSWYQAGSAR SGPVTVYPYM HALDTHHHHH H
Greater than 95.0% as determined by SDS-PAGE.
1 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
IGSF8 protein solution (0.5 mg/ml) contains Phosphate Buffered Saline (pH 7.4) and 30% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers