missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IGSF8 Protein
A cDNA sequence encoding the IGSF8 was constructed and used to recombinantly synthesize the protein.
131.00 € - 1740.00 €
Specifications
Name | IGSF8 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15960259
|
enQuireBio™
QP12389-1UG |
1 μg |
131.00 €
1µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15980259
|
enQuireBio™
QP12389-5UG |
5 μg |
203.00 €
5µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15970259
|
enQuireBio™
QP12389-50UG |
50 μg |
1740.00 €
50µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
IGSF8 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
IGSF8 protein solution (0.5 mg/ml) contains Phosphate Buffered Saline (pH 7.4) and 30% glycerol. |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
ADLREVLVPE GPLYRVAGTA VSISCNVTGY EGPAQQNFEW FLYRPEAPDT ALGIVSTKDT QFSYAVFKSR VVAGEVQVQR LQGDAVVLKI ARLQAQDAGI YECHTPSTDT RYLGSYSGKV ELRVLPDVLQ VSAAPPGPRG RQAPTSPPRM TVHEGQELAL GCLARTSTQK HTHLAVSFGR SVPEAPVGRS TLQEVVGIRS DLAVEAGAPY AERLAAGELR LGKEGTDRYR MVVGGAQAGD AGTYHCTAAE WIQDPDGSWA QIAEKRAVLA HVDVQTLSSQ LAVTVGPGER RIGPGEPLEL LCNVSGALPP AGRHAAYSVG WEMAPAGAPG PGRLVAQLDT EGVGSLGPGY EGRHIAMEKV ASRTYRLRLE AARPGDAGTY RCLAKAYVRG SGTRLREAAS ARSRPLPVHV REEGVVLEAV AWLAGGTVYR GETASLLCNI SVRGGPPGLR LAASWWVERP EDGELSSVPA QLVGGVGQDG VAELGVRPGG GPVSVELVGP RSHRLRLHSL GPEDEGVYHC APSAWVQHAD YSWYQAGSAR SGPVTVYPYM HALDTHHHHH H | |
Greater than 95.0% as determined by SDS-PAGE. |