missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IL34 Protein
A cDNA sequence encoding the IL34 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12421-2ug
Additional Details : Weight : 0.01000kg
Specifications
146433 | |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MRGSHHHHHH GMASMTGGQQ MGRDLYDDDD KDRWGSHMNE PLEMWPLTQN EECTVTGFLR DKLQYRSRLQ YMKHYFPINY KISVPYEGVF RIANVTRLQR AQVSERELRY LWVLVSLSAT ESVQDVLLEG HPSWKYLQEV QTLLLNVQQG LTDVEVSPKV ESVLSLLNAP GPNLKLVRPK ALLDNCFRVM ELLYCSCCKQ SSVLNWQDCE VPSPQSCSPE PSLQYAATQL YPPPPWSPSS PPHSTGSVRP VRAQGEGLLP | |
Greater than 85% as determined by SDS-PAGE. |
IL34 Protein | |
Research Use Only | |
IL34 | |
Human | |
His | |
The IL34 solution (1 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 10% glycerol and 0.4M Urea. |