missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IL3RA Protein
A cDNA sequence encoding the IL3RA was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12424-5ug
Additional Details : Weight : 0.01000kg
Specifications
IL3RA Protein | |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
ADLKEDPNPP ITNLRMKAKA QQLTWDLNRN VTDIECVKDA DYSMPAVNNS YCQFGAISLC EVTNYTVRVA NPPFSTWILF PENSGKPWAG AENLTCWIHD VDFLSCSWAV GPGAPADVQY DLYLNVANRR QQYECLHYKT DAQGTRIGCR FDDISRLSSG SQSSHILVRG RSAAFGIPCTDKFVVFSQIE ILTPPNMTAK CNKTHSFMHW KMRSHFNRKF RYELQIQKRM QPVITEQVRD RTSFQLLNPG TYTVQIRARE RVYEFLSAWS TPQRFECDQE EGANTRAWRH HHHHH | |
Greater than 90.0% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
IL3RA protein solution (0.5 mg/ml) contains 10% glycerol and Phosphate buffered saline (pH 7.4). |