missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IL3RA Protein
A cDNA sequence encoding the IL3RA was constructed and used to recombinantly synthesize the protein.
131.00 € - 1740.00 €
Specifications
Name | IL3RA Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15930379
|
enQuireBio™
QP12424-1UG |
1 μg |
131.00 €
1µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15950379
|
enQuireBio™
QP12424-5UG |
5 μg |
203.00 €
5µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15940379
|
enQuireBio™
QP12424-50UG |
50 μg |
1740.00 €
50µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
IL3RA Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
IL3RA protein solution (0.5 mg/ml) contains 10% glycerol and Phosphate buffered saline (pH 7.4). |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
ADLKEDPNPP ITNLRMKAKA QQLTWDLNRN VTDIECVKDA DYSMPAVNNS YCQFGAISLC EVTNYTVRVA NPPFSTWILF PENSGKPWAG AENLTCWIHD VDFLSCSWAV GPGAPADVQY DLYLNVANRR QQYECLHYKT DAQGTRIGCR FDDISRLSSG SQSSHILVRG RSAAFGIPCTDKFVVFSQIE ILTPPNMTAK CNKTHSFMHW KMRSHFNRKF RYELQIQKRM QPVITEQVRD RTSFQLLNPG TYTVQIRARE RVYEFLSAWS TPQRFECDQE EGANTRAWRH HHHHH | |
Greater than 90.0% as determined by SDS-PAGE. |