missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human KLRG1 His Protein

Product Code. 18147241 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.01 mg
0.05 mg
Unit Size:
0.01mg
0.05mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18147241 0.01 mg 0.01mg
18128131 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18147241 Supplier Novus Biologicals™ Supplier No. NBP2231720.01MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A denatured recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 60-189 of Human KLRG1 The Recombinant Human KLRG1 Protein is derived from E. coli. The Recombinant Human KLRG1 Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number NP_005801
For Use With (Application) SDS-PAGE
Formulation 20 mM Tris-HCl buffer (pH 8.0), 0.4 M UREA, 10% glycerol
Gene ID (Entrez) 10219
Molecular Weight (g/mol) 17 kDa
Name KLRG1 Protein
Purification Method SDS-PAGE
Quantity 0.01 mg
Source E. Coli
Immunogen KLRG1, 60-189 aa, Human, His tag, E.coli MGSSHHHHHH SSGLVPRGSH MGSLCQGSNY STCASCPSCP DRWMKYGNHC YYFSVEEKDW NSSLEFCLAR DSHLLVITDN QEMSLLQVFL SEAFCWIGLR NNSGWRWEDG SPLNFSRISS NSFVQTCGAI NKNGLQASSC EVPLHWVCKK VRL
Storage Requirements -20°Cor to +80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Research Category Immunology, Innate Immunity, Signal Transduction
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.