missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human LAMTOR4 Protein
A cDNA sequence encoding the LAMTOR4 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12534-100ug
Additional Details : Weight : 0.01000kg
Specifications
389541 | |
100 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMTSALTQ GLERIPDQLG YLVLSEGAVL ASSGDLENDE QAASAISELV STACGFRLHR GMNVPFKRLS VVFGEHTLLV TVSGQRVFVV KRQNRGREPI DV | |
Greater than 85.0% as determined by SDS-PAGE. |
LAMTOR4 Protein | |
Research Use Only | |
LAMTOR4 | |
Human | |
His | |
LAMTOR4 protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 10% glycerol and 1mM DTT. |