missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human LAMTOR4 Protein
A cDNA sequence encoding the LAMTOR4 was constructed and used to recombinantly synthesize the protein.
131.00 € - 1740.00 €
Specifications
Gene ID (Entrez) | 389541 |
---|---|
Name | LAMTOR4 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | LAMTOR4 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15930739
|
enQuireBio™
QP12534-2UG |
2 μg |
131.00 €
2µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15920739
|
enQuireBio™
QP12534-10UG |
10 μg |
203.00 €
10µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15910739
|
enQuireBio™
QP12534-100UG |
100 μg |
1740.00 €
100µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
389541 | |
Research Use Only | |
LAMTOR4 | |
Human | |
His | |
LAMTOR4 protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 10% glycerol and 1mM DTT. |
LAMTOR4 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMTSALTQ GLERIPDQLG YLVLSEGAVL ASSGDLENDE QAASAISELV STACGFRLHR GMNVPFKRLS VVFGEHTLLV TVSGQRVFVV KRQNRGREPI DV | |
Greater than 85.0% as determined by SDS-PAGE. |