missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human LCAT Protein

Product Code. 18187799 Shop All Bio Techne Products
Click to view available options
Quantity:
0.5 mg
100 μg
Unit Size:
0.50mg
100µg
This item is not returnable. View return policy

Product Code. 18187799

Brand: Novus Biologicals™ NBP253036100UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A denatured recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 25-440 of Human LCAT The Recombinant Human LCAT Protein is derived from E. coli. The Recombinant Human LCAT Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0) containing 0.4M urea, 10% glycerol
Gene ID (Entrez) 3931
Molecular Weight (g/mol) 49.8kDa
Name Human LCAT Protein
Purification Method >85%, by SDS-PAGE
Quantity 100 μg
Source E.Coli
Immunogen MGSSHHHHHH SSGLVPRGSH MGSHMFWLLN VLFPPHTTPK AELSNHTRPV ILVPGCLGNQ LEAKLDKPDV VNWMCYRKTE DFFTIWLDLN MFLPLGVDCW IDNTRVVYNR SSGLVSNAPG VQIRVPGFGK TYSVEYLDSS KLAGYLHTLV QNLVNNGYVR DETVRAAPYD WRLEPGQQEE YYRKLAGLVE EMHAAYGKPV FLIGHSLGCL HLLYFLLRQP QAWKDRFIDG FISLGAPWGG SIKPMLVLAS GDNQGIPIMS SIKLKEEQRI TTTSPWMFPS RMAWPEDHVF ISTPSFNYTG RDFQRFFADL HFEEGWYMWL QSRDLLAGLP APGVEVYCLY GVGLPTPRTY IYDHGFPYTD PVGVLYEDGD DTVATRSTEL CGLWQGRQPQ PVHLLPLHGI QHLNMVFSNL TLEHINAILL GAYRQGPPAS PTASPEPPPP E
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Regulatory Status RUO
Gene Alias EC 2.3.1.43, lecithin-cholesterol acyltransferasePhospholipid-cholesterol acyltransferase, phosphatidylcholine-sterol acyltransferase
Gene Symbol LCAT
Product Type Recombinant Protein
Conjugate Unconjugated
Cross Reactivity Human
Recombinant Recombinant
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.