missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Leptin Protein
Recombinant Protein
Brand: enQuireBio™ QP5350-10mg
Additional Details : Weight : 0.01000kg
Specifications
Leptin Protein | |
Research Use Only | |
LEP | |
Recombinant Protein | |
E. coli | |
MVPIQKVQDD TKTLIKTIVT RINDISHTQS VSSKQKVTGL DFIPGLHPIL TLSKMDQTLA VYQQILTSMP SRNVIQISND LENLRDLLHV LAFSKSCHLP WASGLETLDS LGGVLEASGY STEVVALSRL QGSLQDMLWQ LDLSPGC | |
Greater than 95% as determined by SDS-PAGE. |
10 mg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Bioactivity of Human Leptin was examined and confirmed in a Ba/F3 cell proliferation assay. This product has not been tested for in vivo use. | |
Human | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |