missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Leptin Protein
Recombinant Protein
110.00 € - 650.00 €
Specifications
Name | Leptin Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Less than or equal to 1.00 EUs/ug as determined by the LAL method. |
Gene Symbol | LEP |
Biological Activity | Bioactivity of Human Leptin was examined and confirmed in a Ba/F3 cell proliferation assay. This product has not been tested for in vivo use. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15955949
|
enQuireBio™
QP5350-200UG |
200 μg |
110.00 €
200µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15945949
|
enQuireBio™
QP5350-1MG |
1 mg |
252.00 €
1mg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15965949
|
enQuireBio™
QP5350-5MG |
5 mg |
373.00 €
5mg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15935949
|
enQuireBio™
QP5350-10MG |
10 mg |
650.00 €
10mg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Leptin Protein | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Bioactivity of Human Leptin was examined and confirmed in a Ba/F3 cell proliferation assay. This product has not been tested for in vivo use. | |
Human | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |
Research Use Only | |
LEP | |
Recombinant Protein | |
E. coli | |
MVPIQKVQDD TKTLIKTIVT RINDISHTQS VSSKQKVTGL DFIPGLHPIL TLSKMDQTLA VYQQILTSMP SRNVIQISND LENLRDLLHV LAFSKSCHLP WASGLETLDS LGGVLEASGY STEVVALSRL QGSLQDMLWQ LDLSPGC | |
Greater than 95% as determined by SDS-PAGE. |