missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human LIMS1 Protein
A DNA sequence encoding the Homo sapiens (Human) LIMS1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6936-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001180411.1 | |
LIMS1 Protein | |
Research Use Only | |
LIMS1 | |
Recombinant Protein | |
E. coli | |
MANALASATC ERCKGGFAPA EKIVNSNGEL YHEQCFVCAQ CFQQFPEGLF YEFEGRKYCE HDFQMLFAPC CHQCGEFTIG RVIKAMNNSW HPECFRCDLC QEVLADIGFV KNAGRHLCRP CHNREKARGL GKYICQKCHA IIDEQPLIFK NDPYHPDHFN CANCGKELTA DARELKGELY CLPCHDKMGV PICGACRRPI EGRVVNAMGK QWHVEHFVCA KCEKPFLGHR HYERKGLAYC ETHYNQLFGD VCFHCNRVIE GGVVSALNKA WCVNCFACST CNTKLTLKNK FVEFDMKPVC KKCYEKFPLE LKKRLKKLAE TLGRK | |
Greater than 80% as determined by SDS-PAGE. |
3987 | |
100 μg | |
Not determined. | |
Full Length of BC005341 | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |