missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human LIMS1 Protein
A DNA sequence encoding the Homo sapiens (Human) LIMS1, was expressed in the hosts and tags indicated.
245.00 € - 3260.00 €
Specifications
Accession Number | NP_001180411.1 |
---|---|
Gene ID (Entrez) | 3987 |
Name | LIMS1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16036770
|
enQuireBio™
QP6936-EC-100UG |
100 μg |
735.00 €
100µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16056770
|
enQuireBio™
QP6936-EC-1MG |
1 mg |
3260.00 €
1mg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16046770
|
enQuireBio™
QP6936-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16066770
|
enQuireBio™
QP6936-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16076770
|
enQuireBio™
QP6936-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16086770
|
enQuireBio™
QP6936-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001180411.1 | |
LIMS1 Protein | |
Not determined. | |
Full Length of BC005341 | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
3987 | |
Research Use Only | |
LIMS1 | |
Recombinant Protein | |
E. coli | |
MANALASATC ERCKGGFAPA EKIVNSNGEL YHEQCFVCAQ CFQQFPEGLF YEFEGRKYCE HDFQMLFAPC CHQCGEFTIG RVIKAMNNSW HPECFRCDLC QEVLADIGFV KNAGRHLCRP CHNREKARGL GKYICQKCHA IIDEQPLIFK NDPYHPDHFN CANCGKELTA DARELKGELY CLPCHDKMGV PICGACRRPI EGRVVNAMGK QWHVEHFVCA KCEKPFLGHR HYERKGLAYC ETHYNQLFGD VCFHCNRVIE GGVVSALNKA WCVNCFACST CNTKLTLKNK FVEFDMKPVC KKCYEKFPLE LKKRLKKLAE TLGRK | |
Greater than 80% as determined by SDS-PAGE. |