missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human M-CSF Protein

Product Code. p-200055427 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
500 μg
Unit Size:
100µg
10µg
1mg
500µg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18478021 10 μg 10µg
18450372 1 mg 1mg
18478891 500 μg 500µg
18405172 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 18478021 Supplier Novus Biologicals™ Supplier No. NBP23497510ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

MCSF Protein is fully biologically active when compared to standard. The ED50 as determined by a cell proliferation assay using murine M-NFS-60 cells is less than 1 ng/mL, corresponding to a specific activity of >1.0 x 106IU/mg.
TRUSTED_SUSTAINABILITY

Specifications

Concentration LYOPH
For Use With (Application) PAGE, Bioactivity
Formulation Lyophilized from a 0.2 um filtered solution in PBS, pH7.4.
Gene ID (Entrez) 1435
Molecular Weight (g/mol) 36.8 kDa
Quantity 10 μg
Storage Requirements Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles.
Endotoxin Concentration Less than 1 EU/ug of M-CSF as determined by LAL method.
Gene Alias colony stimulating factor 1 (macrophage), CSF-1, Lanimostim, macrophage colony stimulating factor, macrophage colony-stimulating factor 1, M-CSF, MCSFlanimostim, MGC31930
Gene Symbol CSF1
Reconstitution Recommended to centrifuge prior to opening. Reconstitute in sterile distilled water to a concentration of 0.1mg/mL. Apportion stock solutions into working aliquots and store at <-20°C.
Research Category Biologically Active Proteins, Cell Cycle and Replication, Cytokine Research, Stem Cell Markers
Purity or Quality Grade >95% by SDS-PAGE and HPLC
Protein M-CSF
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.