missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MTDH Protein
A cDNA sequence encoding the MTDH was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12748-10ug
Additional Details : Weight : 0.01000kg
Specifications
MTDH Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MKHHHHHHASVSSGLNENLT VNGGGWNEKS VKLSSQISAG EEKWNSVSPA SAGKRKAEPS AWSQDTGDAN TNGKDWGRSW SDRSIFSGIG STAEPVSQST TSDYQWDVSR NQPYIDDEWS GLNGLSSADP NSDWNAPAEE WGNWVDEERA SLLKSQEPIP DDQKVSDDDK EKGEGALPTG KSKKKKKKKK KQGEDN | |
Greater than 90.0% as determined by SDS-PAGE. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
MTDH was filtered (0.4microm) and lyophilized in 20mM Tris buffer and 50mM NaCl, pH 7.5. |