missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Myosin-6 Protein

A DNA sequence encoding the Homo sapiens (Human) Myosin-6, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9299-ye-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16038041

  • 615.00 € / 100µg
Estimated Shipment: 25-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_002462.2
Myosin-6 Protein
Research Use Only
MYH6
Recombinant Protein
Yeast
NAYQYMLTDR ENQSILITGE SGAGKTVNTK RVIQYFASIA AIGDRGKKDN ANANKGTLED QIIQANPALE AFGNAKTVRN DNSSRFGKFI RIHFGATGKL ASADIETYLL EKSRVIFQLK AERNYHIFYQ ILSNKKPELL DMLLVTNNPY DYAFVSQGEV SVASIDDSEE LMATDSAFDV LGFTSEEKAG VYKLTGAIMH YGNMKFKQKQ REEQAEPDGT EDADKSAYLM GLNSADLLKG LCHPRVKVGN EYVTKGQSVQ QVYYSIGALA KAVYEKMFNW MVTRINATLE TKQPRQYFIG VLDIAGFEIF DFNSFEQLCI NFTNEKLQQF FNHHMFVLEQ EEYKKEGIEW TFIDFGMDLQ ACIDLIEKPM GIMSILEEEC MFPKATDMTF KAKLYDNHLG KSNNFQKPRN IKGKQEAHFS LIHYAGTVDY NILGWLEKNK DPLNETVVAL YQKSSLKLMA TLFSSYATAD TGDSGKSKGG KKKGSSFQTV SALHRENLNK LMTNLRTTHP HFVRCIIPNE RKAPGVMDNP LVMHQLRCNG VLEGIRICRK GFPNRILYGD FRQRYRILNP VAIPEGQFID SRKGTEKLLS SLDIDHNQYK FGHTKVFFKA GLLGLLEEMR DERLSRIITR MQAQARGQLM RIEFKKIVER RDALLVI
Greater than 90% as determined by SDS-PAGE.
4624
100 μg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers