missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Myosin-6 Protein
A DNA sequence encoding the Homo sapiens (Human) Myosin-6, was expressed in the hosts and tags indicated.
202.00 € - 2155.00 €
Specifications
Accession Number | NP_002462.2 |
---|---|
Gene ID (Entrez) | 4624 |
Name | Myosin-6 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16038041
|
enQuireBio™
QP9299-YE-100UG |
100 μg |
615.00 €
100µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16078041
|
enQuireBio™
QP9299-YE-500UG |
500 μg |
1665.00 €
500µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16048041
|
enQuireBio™
QP9299-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16068041
|
enQuireBio™
QP9299-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16088041
|
enQuireBio™
QP9299-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16058041
|
enQuireBio™
QP9299-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_002462.2 | |
Myosin-6 Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
4624 | |
Research Use Only | |
MYH6 | |
Recombinant Protein | |
Yeast | |
NAYQYMLTDR ENQSILITGE SGAGKTVNTK RVIQYFASIA AIGDRGKKDN ANANKGTLED QIIQANPALE AFGNAKTVRN DNSSRFGKFI RIHFGATGKL ASADIETYLL EKSRVIFQLK AERNYHIFYQ ILSNKKPELL DMLLVTNNPY DYAFVSQGEV SVASIDDSEE LMATDSAFDV LGFTSEEKAG VYKLTGAIMH YGNMKFKQKQ REEQAEPDGT EDADKSAYLM GLNSADLLKG LCHPRVKVGN EYVTKGQSVQ QVYYSIGALA KAVYEKMFNW MVTRINATLE TKQPRQYFIG VLDIAGFEIF DFNSFEQLCI NFTNEKLQQF FNHHMFVLEQ EEYKKEGIEW TFIDFGMDLQ ACIDLIEKPM GIMSILEEEC MFPKATDMTF KAKLYDNHLG KSNNFQKPRN IKGKQEAHFS LIHYAGTVDY NILGWLEKNK DPLNETVVAL YQKSSLKLMA TLFSSYATAD TGDSGKSKGG KKKGSSFQTV SALHRENLNK LMTNLRTTHP HFVRCIIPNE RKAPGVMDNP LVMHQLRCNG VLEGIRICRK GFPNRILYGD FRQRYRILNP VAIPEGQFID SRKGTEKLLS SLDIDHNQYK FGHTKVFFKA GLLGLLEEMR DERLSRIITR MQAQARGQLM RIEFKKIVER RDALLVI | |
Greater than 90% as determined by SDS-PAGE. |