missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NDUFS6 Protein
A cDNA sequence encoding the NDUFS6 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12821-1mg
Additional Details : Weight : 0.01000kg
Specifications
4726 | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSFGVRVSP TGEKVTHTGQ VYDDKDYRRI RFVGRQKEVN ENFAIDLIAE QPVSEVETRV IACDGGGGAL GHPKVYINLD KETKTGTCGY CGLQFRQHHH | |
Greater than 90% as determined by SDS-PAGE. |
NDUFS6 Protein | |
Research Use Only | |
NDUFS6 | |
Human | |
Untagged | |
The NDUFS6 solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 1mM DTT and 30% glycerol. |