missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NDUFS6 Protein
A cDNA sequence encoding the NDUFS6 was constructed and used to recombinantly synthesize the protein.
131.00 € - 3915.00 €
Specifications
Gene ID (Entrez) | 4726 |
---|---|
Name | NDUFS6 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | NDUFS6 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15931659
|
enQuireBio™
QP12821-5UG |
5 μg |
131.00 €
5µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15921659
|
enQuireBio™
QP12821-20UG |
20 μg |
203.00 €
20µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15911659
|
enQuireBio™
QP12821-1MG |
1 mg |
3915.00 €
1mg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
4726 | |
Research Use Only | |
NDUFS6 | |
Human | |
Untagged | |
The NDUFS6 solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 1mM DTT and 30% glycerol. |
NDUFS6 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSFGVRVSP TGEKVTHTGQ VYDDKDYRRI RFVGRQKEVN ENFAIDLIAE QPVSEVETRV IACDGGGGAL GHPKVYINLD KETKTGTCGY CGLQFRQHHH | |
Greater than 90% as determined by SDS-PAGE. |