missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human NDUFS6 Protein

A cDNA sequence encoding the NDUFS6 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12821-5ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15931659

  • 131.00 € / 5µg
Estimated Shipment: 24-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

4726
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSFGVRVSP TGEKVTHTGQ VYDDKDYRRI RFVGRQKEVN ENFAIDLIAE QPVSEVETRV IACDGGGGAL GHPKVYINLD KETKTGTCGY CGLQFRQHHH
Greater than 90% as determined by SDS-PAGE.
NDUFS6 Protein
Research Use Only
NDUFS6
Human
Untagged
The NDUFS6 solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 1mM DTT and 30% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers