missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human Nesfatin-1/Nucleobindin-2 Protein

Product Code. 18492392 Shop All Bio Techne Products
Click to view available options
Quantity:
1 mg
100 μg
500 μg
Unit Size:
100µg
1mg
500µg
This item is not returnable. View return policy

Product Code. 18492392

Brand: Novus Biologicals™ NBP235072100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Nucleobindin 2 Protein is fully biologically active when compared to standard. The biological activity is tested by in vivo assay using healthy wild type male mice (C57BL/6J).
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P80303
Concentration LYOPH
For Use With (Application) PAGE, Bioactivity
Formulation Lyophilized from a 0.2 um filtered concentrated solution in PBS, pH 7.4.
Gene ID (Entrez) 4925
Molecular Weight (g/mol) 9.6 kDa
Quantity 100 μg
Storage Requirements Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles.
Endotoxin Concentration Less than 1 EU/ug of Nesfatin-1/Nucleobindin-2 as determined by LAL method.
Gene Alias DNA-binding protein NEFA, NEFAGastric cancer antigen Zg4, Nesfatin, Nesfatin 1, Nesfatin-1, nucleobindin 2, nucleobindin-2, nucleobinding 2
Gene Symbol NUCB2
Reconstitution Recommended to centrifuge prior to opening. Reconstitute in sterile distilled water or aqueous buffer containing 0.1% BSA to a concentration of 0.1-1.0mg/mL. Apportion stock solutions into working aliquots and store at <-20°C.
Research Category Signal Transduction
Purity or Quality Grade >95% by SDS-PAGE and HPLC
Protein Nesfatin-1/Nucleobindin-2
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.