missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human Nesfatin-1/Nucleobindin-2 Protein
Shop All Bio Techne Products
Click to view available options
Quantity:
1 mg
100 μg
500 μg
Unit Size:
100µg
1mg
500µg
Description
Nucleobindin 2 Protein is fully biologically active when compared to standard. The biological activity is tested by in vivo assay using healthy wild type male mice (C57BL/6J).
Specifications
Specifications
| Accession Number | P80303 |
| Concentration | LYOPH |
| For Use With (Application) | PAGE, Bioactivity |
| Formulation | Lyophilized from a 0.2 um filtered concentrated solution in PBS, pH 7.4. |
| Gene ID (Entrez) | 4925 |
| Molecular Weight (g/mol) | 9.6 kDa |
| Quantity | 100 μg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. |
| Endotoxin Concentration | Less than 1 EU/ug of Nesfatin-1/Nucleobindin-2 as determined by LAL method. |
| Gene Alias | DNA-binding protein NEFA, NEFAGastric cancer antigen Zg4, Nesfatin, Nesfatin 1, Nesfatin-1, nucleobindin 2, nucleobindin-2, nucleobinding 2 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction