missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NEURL2 Protein
A cDNA sequence encoding the NEURL2 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12831-2ug
Additional Details : Weight : 0.01000kg
Specifications
NEURL2 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMAAASEP VDSGALWGLE RPEPPPTRFH RVHGANIRVD PSGTRATRVE SFAHGVCFSR EPLAPGQVFL VEIEEKELGW CGHLRLGLTA LDPASLAPVP EFSLPDLVNL GHTWVFAITR HHNRVPREGR PEAEAAAPSR PPTLLVEPYL RIEQFRIPRDRLVGRSRPGL YSHLLDQLYE LNVLPPTARR SRLGVLFCPR PDGTADMHII INGEDMGPSA RGLPAAQPLY AVVDVFASTK SVRLVQLEYG LPSLQTLCRL VIQRSMVHRL AIDGLHLPKE LKDFCKYE | |
Greater than 85.0% as determined by SDS-PAGE. |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The NEURL2 solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.4M Urea and 10% glycerol. |