missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human NPHS2 Protein

A cDNA sequence encoding the NPHS2 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12869-1mg


Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15961799

  • 7540.00 € / 1mg
Estimated Shipment: 25-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NPHS2 Protein
Research Use Only
Recombinant Protein
E. coli
MKHHHHHHASVKVVQEYERV IIFRLGHLLP GRAKGPGLFF FLPCLDTYHK VDLRLQTLEI PFHEIVTKDM FIMEIDAICY YRMENASLLL SSLAHVSKAV QFLVQTTMKR LLAHRSLTEI LLERKSIAQD AKVALDSVTC IWGIKVERIE IKDVRLPAGL QHSLAVEAEA QRQAKVRMIA AEAEKAASES LRMAAEILSG TPAAVQLRYL HTLQSLSTEK PSTVVLPLPF DLLNCLSSPS NRTQGSLPFP SPSKPVEPLN PKKKDSPML
Greater than 95.0% as determined by SDS-PAGE.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
NPHS2 was filtered (0.4microm) and lyophilized in 30mM acetate buffer and 1mM dithiothreitol, pH 4.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers