missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NPHS2 Protein
A cDNA sequence encoding the NPHS2 was constructed and used to recombinantly synthesize the protein.
131.00 € - 7540.00 €
Specifications
Name | NPHS2 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15971799
|
enQuireBio™
QP12869-2UG |
2 μg |
131.00 €
2µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15951799
|
enQuireBio™
QP12869-10UG |
10 μg |
203.00 €
10µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15961799
|
enQuireBio™
QP12869-1MG |
1 mg |
7540.00 €
1mg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
NPHS2 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
NPHS2 was filtered (0.4microm) and lyophilized in 30mM acetate buffer and 1mM dithiothreitol, pH 4. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MKHHHHHHASVKVVQEYERV IIFRLGHLLP GRAKGPGLFF FLPCLDTYHK VDLRLQTLEI PFHEIVTKDM FIMEIDAICY YRMENASLLL SSLAHVSKAV QFLVQTTMKR LLAHRSLTEI LLERKSIAQD AKVALDSVTC IWGIKVERIE IKDVRLPAGL QHSLAVEAEA QRQAKVRMIA AEAEKAASES LRMAAEILSG TPAAVQLRYL HTLQSLSTEK PSTVVLPLPF DLLNCLSSPS NRTQGSLPFP SPSKPVEPLN PKKKDSPML | |
Greater than 95.0% as determined by SDS-PAGE. |