missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NPHS2 Protein
A cDNA sequence encoding the NPHS2 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12869-2ug
This item is not returnable.
View return policy
Specifications
NPHS2 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MKHHHHHHASVKVVQEYERV IIFRLGHLLP GRAKGPGLFF FLPCLDTYHK VDLRLQTLEI PFHEIVTKDM FIMEIDAICY YRMENASLLL SSLAHVSKAV QFLVQTTMKR LLAHRSLTEI LLERKSIAQD AKVALDSVTC IWGIKVERIE IKDVRLPAGL QHSLAVEAEA QRQAKVRMIA AEAEKAASES LRMAAEILSG TPAAVQLRYL HTLQSLSTEK PSTVVLPLPF DLLNCLSSPS NRTQGSLPFP SPSKPVEPLN PKKKDSPML | |
Greater than 95.0% as determined by SDS-PAGE. |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
NPHS2 was filtered (0.4microm) and lyophilized in 30mM acetate buffer and 1mM dithiothreitol, pH 4. |