missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Nucleolin Protein

A DNA sequence encoding the Homo sapiens (Human) Nucleolin, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9301-ye-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16098041

  • 640.00 € / 100µg
Estimated Shipment: 20-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Nucleolin Protein
Research Use Only
NCL
Recombinant Protein
Yeast
VKLAKAGKNQ GDPKKMAPPP KEVEEDSEDE EMSEDEEDDS SGEEVVIPQK KGKKAAATSA KKVVVSPTKK VAVATPAKKA AVTPGKKAAA TPAKKTVTPA KAVTTPGKKG ATPGKALVAT PGKKGAAIPA KGAKNGKNAK KEDSDEEEDD DSEEDEEDDE DEDEDEDEIE PAAMKAAAAA PASEDEDDED DEDDEDGKSK GIAYIEFKTE ADAEKTFEEK QGTEIDGRSI SLYYTGEKGQ NQDYRGGKNS TWSGESKTLV LSNLSYSATE ETLQEVFEKA TFIKVPQNQN GKSKGYAFIE FASFEDAKEA LNSCNKREIE GRAIRLELQG PRGSPNARSQ PSKTLFVKGL SEDTTEETLK ESFDGSVRAR IVTDRETGSS KGFGFVDFNS EEDAKAAKEA MEDGEIDGNK VTLDWAKPKG EGGFGGRGGG RGGFGGRGGG RGGRGGFGGR GRGGFGGRGG FRGGRGGGGD HKPQGKKTKF E
Greater than 90% as determined by SDS-PAGE.
100 μg
Not determined.
Full Length of Mature Protein
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers