missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Nucleolin Protein
A DNA sequence encoding the Homo sapiens (Human) Nucleolin, was expressed in the hosts and tags indicated.
202.00 € - 2155.00 €
Specifications
Name | Nucleolin Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Gene Symbol | NCL |
Protein Length | Full Length of Mature Protein |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16098041
|
enQuireBio™
QP9301-YE-100UG |
100 μg |
640.00 €
100µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16038051
|
enQuireBio™
QP9301-YE-500UG |
500 μg |
1665.00 €
500µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16008051
|
enQuireBio™
QP9301-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16028051
|
enQuireBio™
QP9301-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16048051
|
enQuireBio™
QP9301-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16018051
|
enQuireBio™
QP9301-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
Nucleolin Protein | |
Not determined. | |
Full Length of Mature Protein | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
Research Use Only | |
NCL | |
Recombinant Protein | |
Yeast | |
VKLAKAGKNQ GDPKKMAPPP KEVEEDSEDE EMSEDEEDDS SGEEVVIPQK KGKKAAATSA KKVVVSPTKK VAVATPAKKA AVTPGKKAAA TPAKKTVTPA KAVTTPGKKG ATPGKALVAT PGKKGAAIPA KGAKNGKNAK KEDSDEEEDD DSEEDEEDDE DEDEDEDEIE PAAMKAAAAA PASEDEDDED DEDDEDGKSK GIAYIEFKTE ADAEKTFEEK QGTEIDGRSI SLYYTGEKGQ NQDYRGGKNS TWSGESKTLV LSNLSYSATE ETLQEVFEKA TFIKVPQNQN GKSKGYAFIE FASFEDAKEA LNSCNKREIE GRAIRLELQG PRGSPNARSQ PSKTLFVKGL SEDTTEETLK ESFDGSVRAR IVTDRETGSS KGFGFVDFNS EEDAKAAKEA MEDGEIDGNK VTLDWAKPKG EGGFGGRGGG RGGFGGRGGG RGGRGGFGGR GRGGFGGRGG FRGGRGGGGD HKPQGKKTKF E | |
Greater than 90% as determined by SDS-PAGE. |