missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human Ornithine Carbamoyltransferase His Protein
Shop All Bio Techne Products
Click to view available options
Unit Size:
250µg
50µg
Description
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 33-354 of Human Ornithine Carbamoyltransferase The Recombinant Human Ornithine Carbamoyltransferase Protein is derived from E. coli. The Recombinant Human Ornithine Carbamoyltransferase Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
| Accession Number | P00480 |
| For Use With (Application) | SDS-PAGE |
| Formulation | Liquid. In 20 mM MES buffer (pH 6.0) containing 2 mM DTT, 10% glycerol, 100 mM NaCl |
| Gene ID (Entrez) | 5009 |
| Molecular Weight (g/mol) | 38.9 kDa |
| Name | Ornithine Carbamoyltransferase Protein |
| Purification Method | SDS-PAGE |
| Storage Requirements | 4°C short term, -20°C long term. Avoid freeze-thaw cycles. |
| Cross Reactivity | Human |
| Purity or Quality Grade | >90% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction