missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human Osteoactivin/GPNMB His Protein
Shop All Bio Techne Products
Click to view available options
Quantity:
250 μg
50 μg
Unit Size:
250µg
50µg
Description
A recombinant protein with a C-Terminal His-tag and corresponding to the amino acids 22-474 of Human Osteoactivin/GPNMB The Recombinant Human Osteoactivin/GPNMB Protein is derived from Baculovirus. The Recombinant Human Osteoactivin/GPNMB Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
| Concentration | 0.5mg/mL |
| For Use With (Application) | SDS-PAGE |
| Formulation | Liquid. PBS (pH 7.4) containing 10% glycerol |
| Gene ID (Entrez) | 10457 |
| Molecular Weight (g/mol) | 51.8 kDa |
| Name | Recombinant Human Osteoactivin/GPNMB Protein |
| Purification Method | >90%, by SDS-PAGE |
| Quantity | 250 μg |
| Source | Baculovirus |
| Storage Requirements | Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles. |
| Show More |
For Research Use Only.
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion