missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human p70 S6 Kinase/S6K Isoform a Protein

Product Code. p-7145930 Shop All Bio Techne Products
Click to view available options
Quantity:
10 μg
50 μg
Unit Size:
10µg
50µg
This item is not returnable. View return policy

Product Code. 18249751

Brand: Novus Biologicals™ NBP26198910UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A recombinant protein with a C-Terminal His-tag and corresponding to the amino acids 1-525 of Human p70 S6 Kinase/S6K The Recombinant Human p70 S6 Kinase/S6K Isoform a Protein is derived from Baculovirus. The Recombinant Human p70 S6 Kinase/S6K Isoform a Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY

Specifications

Concentration 0.25 mg/ml
For Use With (Application) SDS-PAGE
Formulation 50mM Tris-HCl (pH 7.5) containing 30% glycerol, 0.5M NaCl, 2mM DTT, 0.1mM PMSF
Gene ID (Entrez) 6198
Molecular Weight (g/mol) 60.2 kDa
Quantity 10 μg
Source Baculovirus
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Regulatory Status RUO
Endotoxin Concentration < 1.0 EU per 1μg of protein (determined by LAL method)
Gene Symbol RPS6KB1
Cross Reactivity p70 S6 Kinase/S6K
Species Human
Purity or Quality Grade >85%, by SDS-PAGE
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.