missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Parathyroid hormone 2 receptor Protein
A DNA sequence encoding the Homo sapiens (Human) Parathyroid hormone 2 receptor, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6552-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001296445.1 | |
Parathyroid hormone 2 receptor Protein | |
100 μg | |
Not determined. | |
Extracellular Domain | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
5746 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
PTH2R | |
Recombinant Protein | |
E. coli | |
DSDGTITIEE QIVLVLKAKV QCELNITAQL QEGEGNCFPE WDGLICWPRG TVGKISAVPC PPYIYDFNHK GVAFRHCNPN GTWDFMHSLN KTWANYSDCL RFLQPDISIG KQEFFERLY |