missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PDZ and LIM domain protein 1
A DNA sequence encoding the Homo sapiens (Human) PDZ and LIM domain protein 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6480-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_066272.1 | |
PDZ and LIM domain protein 1 | |
Research Use Only | |
PDLIM1 | |
Recombinant Protein | |
E. coli | |
TTQQIDLQGP GPWGFRLVGG KDFEQPLAIS RVTPGSKAAL ANLCIGDVIT AIDGENTSNM THLEAQNRIK GCTDNLTLTV ARSEHKVWSP LVTEEGKRHP YKMNLASEPQ EVLHIGSAHN RSAMPFTASP ASSTTARVIT NQYNNPAGLY SSENISNFNN ALESKTAASG VEANSRPLDH AQPPSSLVID KESEVYKMLQ EKQELNEPPK QSTSFLVLQE ILESEEKGDP NKPSGFRSVK APVTKVAASI GNAQKLPMCD KCGTGIVGVF VKLRDRHRHP ECYVCTDCGT NLKQKGHFFV EDQIYCEKHA RERVTPPEGY EVVTVFPK | |
Greater than 80% as determined by SDS-PAGE. |
9124 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |