missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PHF13 Protein

A cDNA sequence encoding the PHF13 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13038-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15932319

  • 1465.00 € / 50µg
Estimated Shipment: 12-01-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

PHF13 Protein
50 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The PHF13 solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.2M NaCl, 50% glycerol, 2mM DTT, 1mM EDTA and 250mM Imidazole.
Greater than 85% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMDSDSCA AAFHPEEYSP SCKRRRTVED FNKFCTFVLA YAGYIPYPKE ELPLRSSPSP ANSTAGTIDS DGWDAGFSDI ASSVPLPVSD RCFSHLQPTL LQRAKPSNFL LDRKKTDKLK KKKKRKRRDS DAPGKEGYRG GLLKLEAADP YVETPTSPTL QDIPQAPSDP CSGWDSDTPS SGSCATVSPD QVKEIKTEGK RTIVRQGKQV VFRDEDSTGN DEDIMVDSDD DSWDLVTCFC MKPFAGRPMI ECNECHTWIH LSCAKIRKSN VPEVFVCQKC RDSKFDIRRS NRSRTGSRKL FLD
Videos
SDS
Documents
Certificates
Special Offers

Special Offers