missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human PPP2R2B GST (N-Term) Protein

Product Code. 18645090 Shop All Bio Techne Products
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
This item is not returnable. View return policy

Product Code. 18645090

Brand: Novus Biologicals™ H00005521P0210ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Western Blot, ELISA, SDS-PAGE, Protein Array, Immunoaffinity Purification
Formulation 50 mM Tris-HCl, 10 mM reduced Glutathione, pH 8.0 in the elution buffer.
Gene ID (Entrez) 5521
Molecular Weight (g/mol) MolecularWeight-theroretical: 78.1 kDa
Quantity 10 μg
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Gene Alias B55BETA, FLJ95686, MGC24888, PP2A subunit B isoform B55-beta, PP2A subunit B isoform beta, PP2A subunit B isoform PR55-beta, PP2A subunit B isoform R2-beta, PP2A, subunit B, B-beta isoform, PP2AB55BETA, PP2ABBETA, PP2APR55B, PP2APR55BETA, PR2AB55BETA, PR2ABBETA, PR2APR55BETA, PR52B, PR55BETA, PR55-BETA, protein phosphatase 2 (formerly 2A), regulatory subunit B (PR 52), beta isoform, protein phosphatase 2 (formerly 2A), regulatory subunit B, beta isoform, protein phosphatase 2, regulatory subunit B, beta, SCA12, serine/threonine protein phosphatase 2A, 55 kDa regulatory subunit B, betaisoform, serine/threonine protein phosphatase 2A, neuronal isoform, serine/threonine-protein phosphatase 2A 55 kDa regulatory subunit B betaisoform, spinocerebellar ataxia 12
Gene Symbol PPP2R2B
Research Category Stem Cell Markers
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Protein PPP2R2B
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.